Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190453.1 | NP_001177382.1 | 13 - 14 | 1276 | 67 | |
NM_001190454.1 | NP_001177383.1 | 12 - 13 | 1285 | 67 | ||
NM_027151.2 | NP_081427.1 | 12 - 13 | 1270 | 67 | ||
GenBank mRNA | AK144464.1 | - | 12 - 13 | 1146 | 67 | |
AK144505.1 | - | 12 - 13 | 1147 | 67 | ||
AK164021.1 | - | 12 - 13 | 1147 | 67 | ||
AK180539.1 | - | 3 - 4 | 244 | 67 | ||
BC004613.1 | - | 12 - 13 | 1133 | 3585310 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |