Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027352.4 | NP_081628.3 | 3 - 4 | 532 | 134 | |
NR_027343.1 | 4 - 5 | 729 | 134 | |||
GenBank mRNA | AK002515.1 | - | 4 - 5 | 611 | 134 | |
AK010664.2 | - | 3 - 4 | 451 | 134 | ||
AK033413.1 | - | 3 - 4 | 466 | 134 | ||
AK044691.1 | - | 417 | 134 | |||
AK077153.1 | - | 3 - 4 | 408 | 133 | ||
AK077683.1 | - | 3 - 4 | 467 | 134 | ||
AK077706.1 | - | 3 - 4 | 399 | 134 | ||
AK088239.1 | - | 3 - 4 | 413 | 134 | ||
AK148337.1 | - | 3 - 4 | 466 | 134 | ||
AK151843.1 | - | 3 - 4 | 414 | 134 | ||
AK152475.1 | - | 3 - 4 | 414 | 134 | ||
AK152544.1 | - | 3 - 4 | 414 | 134 | ||
AK152603.1 | - | 3 - 4 | 442 | 134 | ||
AK153049.1 | - | 3 - 4 | 414 | 134 | ||
AK153555.1 | - | 3 - 4 | 465 | 134 | ||
AK159469.1 | - | 3 - 4 | 464 | 134 | ||
AK167823.1 | - | 3 - 4 | 466 | 134 | ||
AK168928.1 | - | 4 - 5 | 662 | 134 | ||
AK169382.1 | - | 3 - 4 | 414 | 134 | ||
AK216052.1 | - | 1 - 2 | 71 | 134 | ||
BC005600.1 | - | 3 - 4 | 405 | 3482776 | 134 | |
BC016455.1 | - | 2 - 3 | 257 | 4234343 | 134 | |
BC024982.1 | - | 3 - 4 | 418 | 134 | ||
BU707592.1 | - | 3 - 4 | 532 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |