Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027436.3 | NP_081712.2 | 12 - 13 | 1455 | 109 | |
NR_040642.1 | 12 - 13 | 1455 | 109 | |||
GenBank mRNA | AK017491.1 | - | 12 - 13 | 1431 | 109 | |
AK038479.1 | - | 12 - 13 | 1449 | 109 | ||
AK044460.1 | - | 12 - 13 | 1440 | 109 | ||
AK076613.1 | - | 12 - 13 | 1385 | 109 | ||
BC049871.1 | - | 3 - 4 | 295 | 109 | ||
BC110358.1 | - | 12 - 13 | 1433 | 5322591 | 109 | |
BC144798.1 | - | 12 - 13 | 1436 | 109 | ||
BC145711.1 | - | 12 - 13 | 1436 | 40131014 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |