Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027902.2 | NP_082178.2 | 1 - 2 | 215 | 72 | |
GenBank mRNA | AK004939.2 | - | 1 - 2 | 62 | 72 | |
AY240929.1 | - | 1 - 2 | 192 | 72 | ||
BB872026.1 | - | 1 - 2 | 216 | 72 | ||
BC029645.1 | - | 1 - 2 | 192 | 4195486 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |