Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028041.2 | NP_082317.1 | 17 - 18 | 2174 | 95 | |
GenBank mRNA | AF319547.1 | - | 17 - 18 | 2534 | 95 | |
AK010484.1 | - | 14 - 15 | 1755 | 95 | ||
AK028274.1 | - | 17 - 18 | 2189 | 95 | ||
AK031534.1 | - | 17 - 18 | 2184 | 95 | ||
AK031679.1 | - | 17 - 18 | 2192 | 95 | ||
AK147715.1 | - | 17 - 18 | 2190 | 95 | ||
AK152989.1 | - | 17 - 18 | 2190 | 95 | ||
AK155138.1 | - | 17 - 18 | 2186 | 95 | ||
AK167603.1 | - | 17 - 18 | 2188 | 95 | ||
AK168455.1 | - | 17 - 18 | 2188 | 95 | ||
AK169255.1 | - | 17 - 18 | 2187 | 95 | ||
AK170967.1 | - | 17 - 18 | 2186 | 95 | ||
BC021900.1 | - | 8 - 9 | 1179 | 95 | ||
BC043699.1 | - | 17 - 18 | 2174 | 5344158 | 95 | |
BC055026.1 | - | 13 - 14 | 1617 | 95 | ||
BC071251.1 | - | 11 - 12 | 1471 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |