Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028123.3 | NP_082399.3 | 2 - 3 | 246 | 105 | |
GenBank mRNA | AK076213.1 | - | 2 - 3 | 189 | 105 | |
AK077836.1 | - | 2 - 3 | 190 | 105 | ||
AK145242.1 | - | 2 - 3 | 215 | 105 | ||
AK167716.1 | - | 2 - 3 | 202 | 105 | ||
BC005744.1 | - | 2 - 3 | 211 | 105 | ||
BC099673.1 | - | 2 - 3 | 183 | 4936456 | 105 | |
BC100391.1 | - | 4 - 5 | 426 | 105 | ||
CJ161523.1 | - | 2 - 3 | 247 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |