Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900005755.mm.1 |
NM_028283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005755.mm.1 |
NM_028283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005755.mm.1 |
TC0900000769.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005756.mm.1 |
NM_028283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005756.mm.1 |
NM_028283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005756.mm.1 |
TC0900000769.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005754.mm.1 |
NM_028283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005754.mm.1 |
NM_028283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005754.mm.1 |
TC0900000769.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005758.mm.1 |
NM_028283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005758.mm.1 |
NM_028283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005758.mm.1 |
TC0900000769.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005757.mm.1 |
NM_028283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005757.mm.1 |
NM_028283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005757.mm.1 |
TC0900000769.mm.1 |
MTA TC |