Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028665.4 | NP_082941.2 | 7 - 8 | 1872 | 95 | |
GenBank mRNA | AK016847.1 | - | 2 - 3 | 153 | 95 | |
AK029511.1 | - | 7 - 8 | 1874 | 95 | ||
AK029888.1 | - | 7 - 8 | 1833 | 95 | ||
AK033599.1 | - | 7 - 8 | 1861 | 95 | ||
AK048925.1 | - | 7 - 8 | 1766 | 95 | ||
AK133339.1 | - | 1833 | 95 | |||
BC030885.1 | - | 7 - 8 | 1501 | 95 | ||
BC100433.1 | - | 7 - 8 | 1908 | 95 | ||
BC145456.1 | - | 7 - 8 | 990 | 9054017 | 95 | |
BC145457.1 | - | 6 - 7 | 854 | 95 | ||
BC145458.1 | - | 6 - 7 | 854 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |