Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001326.1 | NP_001001326.1 | 16 - 17 | 3052 | 82 | |
NM_029811.2 | NP_084087.2 | 15 - 16 | 1801 | 82 | ||
NR_037617.1 | 16 - 17 | 3233 | 82 | |||
GenBank mRNA | AK008100.1 | - | 8 - 9 | 875 | 82 | |
BC030391.1 | - | 12 - 13 | 1527 | 4950637 | 82 | |
BC032266.1 | - | 8 - 9 | 947 | 82 | ||
BC051034.1 | - | 16 - 17 | 3233 | 5345299 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |