Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254762.1 | NP_001241691.1 | 2 - 3 | 516 | 102 | |
NM_030037.2 | NP_084313.1 | 3 - 4 | 669 | 102 | ||
GenBank mRNA | AK088731.1 | - | 2 - 3 | 507 | 102 | |
BC003880.1 | - | 2 - 3 | 526 | 3496491 | 102 | |
BC034167.1 | - | 3 - 4 | 654 | 102 | ||
BC039620.1 | - | 3 - 4 | 650 | 5051528 | 102 | |
BE861793.1 | - | 2 - 3 | 143 | 102 | ||
BQ445267.1 | - | 1 - 2 | 82 | 102 | ||
CK617131.1 | - | 2 - 3 | 467 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |