Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146013.1 | NP_001139485.1 | 4 - 5 | 899 | 82 | |
NM_148931.3 | NP_683733.2 | 3 - 4 | 817 | 82 | ||
GenBank mRNA | AB118159.1 | - | 4 - 5 | 846 | 82 | |
AF411042.3 | - | 4 - 5 | 953 | 82 | ||
AY147186.2 | - | 4 - 5 | 1052 | 82 | ||
BC138125.1 | - | 3 - 4 | 820 | 8861144 | 82 | |
BC138126.1 | - | 3 - 4 | 820 | 8861145 | 82 | |
BC157997.1 | - | 4 - 5 | 899 | 9007308 | 82 | |
BC172125.1 | - | 3 - 4 | 760 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |