Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172258.3 | NP_758462.2 | 9 - 10 | 1245 | 67 | |
GenBank mRNA | AF453745.1 | - | 9 - 10 | 1245 | 67 | |
AK132910.1 | - | 10 - 11 | 1331 | 67 | ||
AY211261.1 | - | 9 - 10 | 1227 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |