Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172289.4 | NP_758493.2 | 2 - 3 | 191 | 72 | |
GenBank mRNA | AF453746.1 | - | 2 - 3 | 190 | 72 | |
AK032074.1 | - | 2 - 3 | 192 | 72 | ||
AK052277.1 | - | 2 - 3 | 192 | 72 | ||
AK150167.1 | - | 2 - 3 | 223 | 72 | ||
BC115964.1 | - | 2 - 3 | 192 | 40057255 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |