Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172289.4 | NP_758493.2 | 8 - 9 | 874 | 73 | |
GenBank mRNA | AF453746.1 | - | 9 - 10 | 873 | 73 | |
AK032074.1 | - | 8 - 9 | 875 | 73 | ||
AK052277.1 | - | 8 - 9 | 875 | 73 | ||
AK150167.1 | - | 8 - 9 | 906 | 73 | ||
BC115964.1 | - | 8 - 9 | 875 | 40057255 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |