Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136471.1 | NP_001129943.1 | 8 - 9 | 1037 | 50 | |
NM_172297.1 | NP_758501.2 | 7 - 8 | 1054 | 50 | ||
GenBank mRNA | AK134892.1 | - | 8 - 9 | 1023 | 50 | |
BC021915.1 | - | 7 - 8 | 1054 | 4527178 | 50 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |