Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172426.2 | NP_766014.1 | 4 - 5 | 1451 | 91 | |
GenBank mRNA | AK012771.1 | - | 257 | 91 | ||
AK083127.1 | - | 4 - 5 | 1335 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mus musculus 1700 Inventoried Probe spans exons |