Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110240.1 | NP_001103710.1 | 8 - 9 | 1862 | 92 | |
NM_172426.2 | NP_766014.1 | 9 - 10 | 1952 | 92 | ||
GenBank mRNA | AK012771.1 | - | 758 | 92 | ||
AK083127.1 | - | 9 - 10 | 1836 | 92 | ||
BC115867.1 | - | 8 - 9 | 1864 | 40081382 | 92 | |
BC116637.1 | - | 7 - 8 | 1678 | 40081381 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |