Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172436.3 | NP_766024.1 | 15 - 16 | 1618 | 101 | |
GenBank mRNA | AK019150.1 | - | 15 - 16 | 1621 | 101 | |
AK030595.1 | - | 15 - 16 | 1648 | 101 | ||
AK083156.1 | - | 15 - 16 | 1619 | 101 | ||
AK144958.1 | - | 15 - 16 | 1652 | 101 | ||
AK147948.1 | - | 15 - 16 | 1634 | 101 | ||
BC013845.1 | - | 15 - 16 | 1597 | 101 | ||
BC024952.1 | - | 15 - 16 | 1811 | 101 | ||
BC060505.1 | - | 15 - 16 | 1605 | 6311836 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |