Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172439.3 | NP_766027.2 | 7 - 8 | 1958 | 76 | |
GenBank mRNA | AK034272.1 | - | 7 - 8 | 1959 | 76 | |
AK136908.1 | - | 7 - 8 | 1945 | 76 | ||
AK165718.1 | - | 6 - 7 | 2486 | 76 | ||
AK170114.1 | - | 7 - 8 | 1924 | 76 | ||
AK171029.1 | - | 7 - 8 | 1919 | 76 | ||
BC022915.1 | - | 7 - 8 | 1911 | 76 | ||
BC046561.1 | - | 6 - 7 | 771 | 76 | ||
BC051452.1 | - | 7 - 8 | 1927 | 76 | ||
BC131634.1 | - | 7 - 8 | 1953 | 40096940 | 76 | |
BC131635.1 | - | 7 - 8 | 1950 | 40096943 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |