Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290684.1 | NP_001277613.1 | 6 - 7 | 3238 | 62 | |
NM_001290686.1 | NP_001277615.1 | 2 - 3 | 370 | 62 | ||
NM_001290687.1 | NP_001277616.1 | 2 - 3 | 552 | 62 | ||
NM_172441.3 | NP_766029.2 | 6 - 7 | 3256 | 62 | ||
GenBank mRNA | AK029338.1 | - | 2 - 3 | 244 | 62 | |
AK032256.1 | - | 2 - 3 | 245 | 62 | ||
AK049850.1 | - | 2 - 3 | 302 | 62 | ||
CF534250.1 | - | 2 - 3 | 513 | 62 | ||
CK635066.1 | - | 3 - 4 | 521 | 62 | ||
CO431718.1 | - | 2 - 3 | 552 | 62 | ||
DQ358971.1 | - | 6 - 7 | 3219 | 62 | ||
EF071946.1 | - | 6 - 7 | 3256 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mus musculus Probe spans exons |