Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172471.2 | NP_766059.1 | 8 - 9 | 1226 | 67 | |
GenBank mRNA | AK087428.1 | - | 8 - 9 | 1154 | 67 | |
AK140756.1 | - | 8 - 9 | 1227 | 67 | ||
BC043314.1 | - | 8 - 9 | 1169 | 5150581 | 67 | |
BC062196.1 | - | 8 - 9 | 1210 | 3490706 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |