Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172471.2 | NP_766059.1 | 9 - 10 | 1538 | 100 | |
GenBank mRNA | AK087428.1 | - | 9 - 10 | 1466 | 100 | |
AK134707.1 | - | 9 - 10 | 1466 | 100 | ||
AK140756.1 | - | 9 - 10 | 1539 | 100 | ||
BC043314.1 | - | 9 - 10 | 1481 | 5150581 | 100 | |
BC062196.1 | - | 9 - 10 | 1522 | 3490706 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |