Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177843.1 | NP_001171314.1 | 13 - 14 | 1548 | 75 | |
NM_001177844.1 | NP_001171315.1 | 2 - 3 | 676 | 75 | ||
NM_172475.3 | NP_766063.3 | 13 - 14 | 1178 | 75 | ||
GenBank mRNA | AK029073.1 | - | 2 - 3 | 655 | 75 | |
AK082365.1 | - | 13 - 14 | 1124 | 75 | ||
AK148414.1 | - | 2 - 3 | 675 | 75 | ||
AK171532.1 | - | 2 - 3 | 604 | 75 | ||
BC058672.1 | - | 13 - 14 | 1169 | 6830456 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |