Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00011521.mm.1 |
NM_001166437.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011521.mm.1 |
NM_001166437.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011521.mm.1 |
NM_172478.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011521.mm.1 |
NM_172478.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011521.mm.1 |
TC0X00001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011522.mm.1 |
NM_001166437.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011522.mm.1 |
NM_001166437.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011522.mm.1 |
NM_172478.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011522.mm.1 |
NM_172478.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011522.mm.1 |
TC0X00001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011525.mm.1 |
NM_172478.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011525.mm.1 |
NM_172478.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011525.mm.1 |
TC0X00001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011524.mm.1 |
NM_172478.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011524.mm.1 |
NM_172478.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011524.mm.1 |
TC0X00001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011526.mm.1 |
NM_001166437.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011526.mm.1 |
NM_001166437.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011526.mm.1 |
NM_172478.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011526.mm.1 |
NM_172478.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011526.mm.1 |
TC0X00001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011523.mm.1 |
NM_172478.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011523.mm.1 |
NM_172478.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011523.mm.1 |
TC0X00001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011527.mm.1 |
NM_001166437.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011527.mm.1 |
NM_001166437.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011527.mm.1 |
NM_172478.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011527.mm.1 |
NM_172478.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011527.mm.1 |
TC0X00001798.mm.1 |
MTA TC |