Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600023233.mm.1 |
TC0600003016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023233.mm.1 |
NM_001172207.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023233.mm.1 |
NM_001172207.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023233.mm.1 |
NM_172492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023233.mm.1 |
NM_172492.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023234.mm.1 |
TC0600003016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023234.mm.1 |
NM_001172207.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023234.mm.1 |
NM_001172207.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023236.mm.1 |
TC0600003016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023236.mm.1 |
NM_172492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023236.mm.1 |
NM_172492.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023231.mm.1 |
TC0600003016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023231.mm.1 |
NM_001172207.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023231.mm.1 |
NM_001172207.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023231.mm.1 |
NM_172492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023231.mm.1 |
NM_172492.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023237.mm.1 |
TC0600003016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023237.mm.1 |
NM_172492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023237.mm.1 |
NM_172492.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023235.mm.1 |
TC0600003016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023235.mm.1 |
NM_172492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023235.mm.1 |
NM_172492.3 |
Refseq |