Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290997.1 | NP_001277926.1 | 16 - 17 | 1951 | 61 | |
NM_001291000.1 | NP_001277929.1 | 12 - 13 | 2069 | 61 | ||
NM_172537.4 | NP_766125.2 | 12 - 13 | 2069 | 61 | ||
NM_199238.3 | NP_954708.1 | 12 - 13 | 2069 | 61 | ||
NM_199239.3 | NP_954709.1 | 11 - 12 | 1304 | 61 | ||
NM_199240.3 | NP_954710.1 | 11 - 12 | 1304 | 61 | ||
NM_199241.3 | NP_954711.1 | 11 - 12 | 1304 | 61 | ||
GenBank mRNA | AB091532.1 | - | 12 - 13 | 2069 | 61 | |
AB091533.1 | - | 11 - 12 | 1250 | 61 | ||
AB091534.1 | - | 11 - 12 | 1250 | 61 | ||
AB091535.1 | - | 11 - 12 | 1250 | 61 | ||
AB091536.1 | - | 11 - 12 | 1250 | 61 | ||
AK031307.1 | - | 12 - 13 | 1765 | 61 | ||
AK052232.1 | - | 12 - 13 | 1477 | 61 | ||
AK084922.1 | - | 12 - 13 | 1766 | 61 | ||
AK122515.1 | - | 16 - 17 | 1954 | 61 | ||
BC060680.1 | - | 12 - 13 | 1363 | 6406252 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |