Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167994.1 | NP_001161466.1 | 11 - 12 | 1361 | 77 | |
NM_172540.2 | NP_766128.2 | 10 - 11 | 1285 | 77 | ||
GenBank mRNA | AK028999.1 | - | 10 - 11 | 1241 | 77 | |
AK049523.1 | - | 10 - 11 | 1283 | 77 | ||
AK129030.1 | - | 10 - 11 | 1478 | 77 | ||
AK137595.1 | - | 1247 | 77 | |||
BC030387.1 | - | 9 - 10 | 946 | 77 | ||
BC056937.1 | - | 11 - 12 | 1302 | 5716144 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |