Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198587.3 | NP_001185516.2 | 15 - 16 | 4122 | 85 | |
NM_001252074.2 | NP_001239003.1 | 2 - 3 | 1337 | 85 | ||
NM_172544.3 | NP_766132.2 | 15 - 16 | 3039 | 85 | ||
GenBank mRNA | AK032126.1 | - | 15 - 16 | 3031 | 85 | |
AK042730.1 | - | 2 - 3 | 1339 | 85 | ||
AK129205.1 | - | 14 - 15 | 2912 | 85 | ||
BC060719.1 | - | 16 - 17 | 3697 | 6406001 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |