Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172554.2 | NP_766142.2 | 10 - 11 | 1177 | 97 | |
GenBank mRNA | AK038670.1 | - | 10 - 11 | 1121 | 97 | |
AK039575.1 | - | 10 - 11 | 1178 | 97 | ||
AK046891.1 | - | 10 - 11 | 1119 | 97 | ||
AK122406.1 | - | 10 - 11 | 1251 | 97 | ||
AK134967.1 | - | 10 - 11 | 1126 | 97 | ||
BC027101.1 | - | 10 - 11 | 1065 | 97 | ||
BC058772.1 | - | 10 - 11 | 1118 | 97 | ||
BC080664.1 | - | 10 - 11 | 1116 | 97 | ||
BC117788.1 | - | 10 - 11 | 1119 | 40092137 | 97 | |
BC117789.2 | - | 10 - 11 | 1119 | 40092140 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |