Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172568.2 | NP_766156.2 | 10 - 11 | 2621 | 70 | |
GenBank mRNA | AK077168.1 | - | 4 - 5 | 1241 | 70 | |
BC117499.1 | - | 4 - 5 | 1235 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |