Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172590.3 | NP_766178.2 | 2 - 3 | 362 | 76 | |
GenBank mRNA | AK046853.1 | - | 2 - 3 | 331 | 76 | |
AK082348.1 | - | 2 - 3 | 363 | 76 | ||
AK149993.1 | - | 2 - 3 | 328 | 76 | ||
AK194741.1 | - | 1 - 2 | 29 | 76 | ||
BB650073.1 | - | 2 - 3 | 358 | 76 | ||
BC138095.1 | - | 2 - 3 | 268 | 8861114 | 76 | |
BC145143.1 | - | 2 - 3 | 264 | 76 | ||
BC145144.1 | - | 2 - 3 | 268 | 9053695 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |