Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172591.3 | NP_766179.2 | 8 - 9 | 1140 | 63 | |
GenBank mRNA | AK031041.1 | - | 8 - 9 | 1143 | 63 | |
AK142282.1 | - | 8 - 9 | 826 | 63 | ||
AK146566.1 | - | 8 - 9 | 874 | 63 | ||
AK164135.1 | - | 8 - 9 | 886 | 63 | ||
BC019933.1 | - | 9 - 10 | 876 | 63 | ||
BC047122.1 | - | 9 - 10 | 876 | 63 | ||
BC052456.1 | - | 8 - 9 | 837 | 6830607 | 63 | |
BC053718.1 | - | 8 - 9 | 856 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |