Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038619.1 | NP_001033708.1 | 19 - 20 | 2487 | 78 | |
NM_172646.2 | NP_766234.1 | 18 - 19 | 2479 | 78 | ||
GenBank mRNA | AK036199.1 | - | 19 - 20 | 2452 | 78 | |
AK049724.1 | - | 18 - 19 | 2480 | 78 | ||
AK082698.1 | - | 19 - 20 | 2431 | 78 | ||
AK147678.1 | - | 19 - 20 | 2431 | 78 | ||
BC004050.1 | - | 7 - 8 | 847 | 78 | ||
BC141143.1 | - | 19 - 20 | 2335 | 9055755 | 78 | |
BC141144.1 | - | 19 - 20 | 2335 | 9055756 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |