Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177627.1 | NP_001171098.1 | 3 - 4 | 551 | 89 | |
NM_172659.2 | NP_766247.2 | 3 - 4 | 551 | 89 | ||
GenBank mRNA | AK079650.1 | - | 3 - 4 | 495 | 89 | |
AK089246.1 | - | 3 - 4 | 543 | 89 | ||
AK137441.1 | - | 3 - 4 | 517 | 89 | ||
AK150104.1 | - | 3 - 4 | 504 | 89 | ||
AK163445.1 | - | 3 - 4 | 512 | 89 | ||
AK167904.1 | - | 3 - 4 | 513 | 89 | ||
AK172411.1 | - | 3 - 4 | 495 | 89 | ||
BC058210.1 | - | 3 - 4 | 475 | 89 | ||
BC141168.1 | - | 3 - 4 | 482 | 9055780 | 89 | |
BC145480.1 | - | 3 - 4 | 482 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |