Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172665.5 | NP_766253.2 | 4 - 5 | 738 | 119 | |
GenBank mRNA | AK038454.1 | - | 3 - 4 | 2253 | 119 | |
AK046805.1 | - | 4 - 5 | 739 | 119 | ||
AK085215.1 | - | 4 - 5 | 637 | 119 | ||
AK153649.1 | - | 4 - 5 | 691 | 119 | ||
AK163553.1 | - | 4 - 5 | 668 | 119 | ||
BC025480.1 | - | 3 - 4 | 479 | 119 | ||
BC027196.1 | - | 4 - 5 | 661 | 4458562 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |