Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pogz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165948.1 | NP_001159420.1 | 7 - 8 | 1224 | 60 | |
NM_172683.3 | NP_766271.2 | 7 - 8 | 1251 | 60 | ||
GenBank mRNA | AK035255.1 | - | 7 - 8 | 1106 | 60 | |
AK161889.1 | - | 7 - 8 | 1079 | 60 | ||
BC065134.1 | - | 7 - 8 | 1240 | 60 | ||
BC066108.1 | - | 7 - 8 | 1240 | 60 | ||
BC085188.1 | - | 7 - 8 | 1084 | 30682742 | 60 | |
BC096492.1 | - | 6 - 7 | 1094 | 30745658 | 60 | |
BC139432.1 | - | 7 - 8 | 1106 | 8862454 | 60 | |
BC139433.1 | - | 7 - 8 | 1106 | 8862455 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |