Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172685.3 | NP_766273.1 | 3 - 4 | 448 | 86 | |
GenBank mRNA | AK032806.1 | - | 3 - 4 | 449 | 86 | |
AK146034.1 | - | 3 - 4 | 447 | 86 | ||
AK170598.1 | - | 3 - 4 | 440 | 86 | ||
AK187917.1 | - | 2 - 3 | 217 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |