Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172695.2 | NP_766283.2 | 9 - 10 | 1640 | 80 | |
GenBank mRNA | AK019070.1 | - | 9 - 10 | 1646 | 80 | |
AK075934.1 | - | 9 - 10 | 1572 | 80 | ||
AK166122.1 | - | 9 - 10 | 1546 | 80 | ||
BC139355.1 | - | 9 - 10 | 1639 | 8862377 | 80 | |
BC139356.1 | - | 9 - 10 | 1639 | 8862378 | 80 | |
BC139773.1 | - | 9 - 10 | 1550 | 40111789 | 80 | |
DV646766.1 | - | 1 - 2 | 126 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |