Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172699.3 | NP_766287.1 | 6 - 7 | 824 | 82 | |
GenBank mRNA | AK082840.1 | - | 6 - 7 | 825 | 82 | |
AK139508.1 | - | 6 - 7 | 758 | 82 | ||
BC058231.1 | - | 6 - 7 | 797 | 6314981 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |