Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162965.1 | NP_001156437.1 | 12 - 13 | 1390 | 66 | |
NM_001254731.1 | NP_001241660.1 | 12 - 13 | 1390 | 66 | ||
NM_172718.3 | NP_766306.2 | 12 - 13 | 1390 | 66 | ||
NR_027934.1 | 3 - 4 | 577 | 66 | |||
NR_027936.1 | 3 - 4 | 577 | 66 | |||
GenBank mRNA | AK010756.1 | - | 3 - 4 | 563 | 66 | |
AK044716.1 | - | 12 - 13 | 1344 | 66 | ||
AK051018.1 | - | 3 - 4 | 578 | 66 | ||
AK053555.1 | - | 12 - 13 | 1324 | 66 | ||
AK167632.1 | - | 3 - 4 | 541 | 66 | ||
AK170021.1 | - | 12 - 13 | 1382 | 66 | ||
BC138049.1 | - | 12 - 13 | 1316 | 8861068 | 66 | |
BC138050.1 | - | 12 - 13 | 1316 | 8861069 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |