Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172735.2 | NP_766323.1 | 1 - 2 | 158 | 81 | |
GenBank mRNA | AJ537495.1 | - | 1 - 2 | 150 | 81 | |
AK088527.1 | - | 1 - 2 | 159 | 81 | ||
AK146228.1 | - | 1 - 2 | 173 | 81 | ||
AK151694.1 | - | 1 - 2 | 154 | 81 | ||
AK164393.1 | - | 1 - 2 | 157 | 81 | ||
AK167465.1 | - | 1 - 2 | 159 | 81 | ||
AK167992.1 | - | 1 - 2 | 173 | 81 | ||
AK168071.1 | - | 1 - 2 | 169 | 81 | ||
BC037445.1 | - | 1 - 2 | 112 | 81 | ||
BC050141.1 | - | 1 - 2 | 151 | 30011212 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |