Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168290.1 | NP_001161762.1 | 6 - 7 | 2837 | 91 | |
NM_172755.3 | NP_766343.3 | 6 - 7 | 2837 | 91 | ||
GenBank mRNA | AF518875.1 | - | 6 - 7 | 2830 | 91 | |
AK042293.1 | - | 6 - 7 | 2838 | 91 | ||
BC023276.1 | - | 2 - 3 | 604 | 91 | ||
BC042763.1 | - | 4 - 5 | 1185 | 91 | ||
BC057305.1 | - | 6 - 7 | 2490 | 6822887 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |