Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168290.1 | NP_001161762.1 | 7 - 8 | 3329 | 75 | |
NM_172755.3 | NP_766343.3 | 7 - 8 | 3329 | 75 | ||
GenBank mRNA | AF518875.1 | - | 7 - 8 | 3322 | 75 | |
AK042293.1 | - | 7 - 8 | 3330 | 75 | ||
BC023276.1 | - | 3 - 4 | 1096 | 75 | ||
BC042763.1 | - | 5 - 6 | 1677 | 75 | ||
BC047383.1 | - | 416 | 75 | |||
BC057305.1 | - | 7 - 8 | 2982 | 6822887 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |