Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172756.2 | NP_766344.2 | 1 - 2 | 90 | 102 | |
GenBank mRNA | AK049395.1 | - | 1 - 2 | 81 | 102 | |
AK078830.1 | - | 1 - 2 | 81 | 102 | ||
AK080768.1 | - | 1 - 2 | 86 | 102 | ||
AK083552.1 | - | 1 - 2 | 73 | 102 | ||
AK087444.1 | - | 1 - 2 | 86 | 102 | ||
BC030436.1 | - | 1 - 2 | 89 | 102 | ||
CJ046325.1 | - | 1 - 2 | 91 | 102 | ||
EU184014.1 | - | 1 - 2 | 66 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |