Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfrkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172766.3 | NP_766354.2 | 15 - 16 | 1688 | 85 | |
GenBank mRNA | AK041206.1 | - | 15 - 16 | 1926 | 85 | |
AK049804.1 | - | 1 - 2 | 63 | 85 | ||
BC033595.1 | - | 15 - 16 | 1678 | 4947357 | 85 | |
BC059861.1 | - | 15 - 16 | 1735 | 85 | ||
BC059900.1 | - | 6 - 7 | 1335 | 85 | ||
BC060076.1 | - | 15 - 16 | 1917 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |