Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172778.2 | NP_766366.2 | 4 - 5 | 479 | 87 | |
GenBank mRNA | AK031833.1 | - | 4 - 5 | 463 | 87 | |
AK054050.1 | - | 4 - 5 | 478 | 87 | ||
BC113182.1 | - | 4 - 5 | 468 | 40061991 | 87 | |
BC113788.1 | - | 4 - 5 | 468 | 40061990 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |