Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172780.3 | NP_766368.2 | 14 - 15 | 1758 | 74 | |
GenBank mRNA | AK032326.1 | - | 14 - 15 | 1739 | 74 | |
AK122232.1 | - | 14 - 15 | 1729 | 74 | ||
BC130221.1 | - | 14 - 15 | 1743 | 40141414 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |