Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172808.2 | NP_766396.2 | 11 - 12 | 1168 | 69 | |
GenBank mRNA | AK077206.1 | - | 11 - 12 | 1169 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |