Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253759.1 | NP_001240688.1 | 18 - 19 | 2513 | 113 | |
NM_172813.3 | NP_766401.1 | 17 - 18 | 2456 | 113 | ||
GenBank mRNA | AK045500.1 | - | 16 - 17 | 2267 | 113 | |
AK051821.1 | - | 17 - 18 | 2404 | 113 | ||
AK082388.1 | - | 15 - 16 | 2293 | 113 | ||
BC052062.1 | - | 15 - 16 | 2024 | 5703279 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |