Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271544.1 | NP_001258473.1 | 3 - 4 | 529 | 68 | |
NM_001271545.1 | NP_001258474.1 | 3 - 4 | 529 | 68 | ||
NM_001271546.1 | NP_001258475.1 | 3 - 4 | 529 | 68 | ||
NM_001271547.1 | NP_001258476.1 | 3 - 4 | 529 | 68 | ||
NM_172830.2 | NP_766418.1 | 3 - 4 | 529 | 68 | ||
GenBank mRNA | AK050187.1 | - | 1 - 2 | 105 | 68 | |
AK085269.1 | - | 3 - 4 | 529 | 68 | ||
AK085424.1 | - | 3 - 4 | 532 | 68 | ||
AK128930.1 | - | 3 - 4 | 531 | 68 | ||
AK142545.1 | - | 3 - 4 | 531 | 68 | ||
AK143939.1 | - | 3 - 4 | 535 | 68 | ||
BC113179.1 | - | 3 - 4 | 524 | 40060674 | 68 | |
BC113785.1 | - | 3 - 4 | 524 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |